missing translation for 'onlineSavingsMsg'
Learn More

N-Iodosuccinimide, MP Biomedicals

Product Code. 11452545 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452545 10 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11452545 Supplier MP Biomedicals Supplier No. 0520850605
  • ONLINE EXCLUSIVE PRICE
    74.95 EUR
    71.40 EUR / 5g
    Save 3.55 EUR
    4% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 516-12-1
Molecular Formula C4H4INO2
MDL Number MFCD00005512
Quantity 10 g
Synonym n-iodosuccinimide, succiniodimide, n-iodo succinimide, 2,5-pyrrolidinedione, 1-iodo, unii-3cos3x3n4p, nis, succinimide, n-iodo, 3cos3x3n4p, 1-iodo-pyrrolidine-2,5-dione, 1-iodoazolidine-2,5-dione
InChI Key LQZMLBORDGWNPD-UHFFFAOYSA-N
SMILES IN1C(=O)CCC1=O
IUPAC Name 1-iodopyrrolidine-2,5-dione
Molecular Weight (g/mol) 224.99
PubChem CID 120273
ChEBI CHEBI:53204
Formula Weight 224.99
Chemical Name or Material N-Iodosuccinimide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.