accessibility menu, dialog, popup

UserName

H3CONNHNN

CAS RN 51449-81-1

IUPAC Name: 5-(2-methoxyphenyl)-2H-1,2,3,4-tetrazole
Synonyms: 5-(2-methoxyphenyl)-2H-1,2,3,4-tetrazole
Molecular Weight (g/mol): 176.18
Molecular Formula: C8H8N4O
InChi Key: ANGNJNYCUCJQJV-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C1=NNN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results