CAS RN 513-53-1
IUPAC Name:
butane-2-thiol
Synonyms:
2-butanethiol
Molecular Weight (g/mol):
90.18
Molecular Formula:
C4H10S
InChi Key:
LOCHFZBWPCLPAN-UHFFFAOYNA-N
SMILES:
CCC(C)S
1
–
4
of
4
results
1
2-Butanethiol, Sigma-Aldrich™
CAS: 513-53-1 Grade: Research Molecular Formula: CH3CH2CH(SH)CH3 Molecular Weight (g/mol): 90.19
2
sec-Butyl Mercaptan, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
sec-Butyl Mercaptan, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
sec-Butyl Mercaptan, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.