Learn More
sec-Butyl Mercaptan, TRC
Specifications
Specifications
| Chemical Name or Material | sec-Butyl Mercaptan |
| CAS | 513-53-1 |
| Molecular Formula | C4H10S |
| Quantity | 10 g |
| Synonym | (±)-2-Butanethiol, 1-Methyl-1-propanethiol, 2-Butyl mercaptan, 2-Butylthiol, 2-Mercaptobutane, 3-Methyl-2-propanethiol, NSC 78417, s-Butyl mercaptan, sec-Butanethiol, sec-Butyl mercaptan, sec-Butyl thioalcohol, sec-Butylthiol, 2-Butanethiol, (±)-2-Butanethiol, 1-Methyl-1-propanethiol, 2-Butyl mercaptan, 2-Butylthiol, 2-Mercaptobutane, 3-Methyl-2-propanethiol, NSC 78417, s-Butyl mercaptan, sec-Butanethiol, sec-Butyl thioalcohol, sec-Butylthiol, 2-Butanethiol |
| InChI Formula | InChI=1S/C4H10S/c1-3-4(2)5/h4-5H,3H2,1-2H3 |
| SMILES | CCC(C)S |
| IUPAC Name | butane-2-thiol |
| Molecular Weight (g/mol) | 90.19 |
| Formula Weight | 90.0503 |