accessibility menu, dialog, popup

UserName

CAS RN 50816-19-8

HOBr

CAS RN 50816-19-8

IUPAC Name: 8-bromooctan-1-ol
Synonyms: 8-bromooctan-1-ol
Molecular Weight (g/mol): 209.13
Molecular Formula: C8H17BrO
InChi Key: GMXIEASXPUEOTG-UHFFFAOYSA-N
SMILES: OCCCCCCCCBr
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)