missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Bromooctan-1-ol, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 8-Bromooctan-1-ol |
| CAS | 50816-19-8 |
| Molecular Formula | C8H17BrO |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C8H17BrO/c9-7-5-3-1-2-4-6-8-10/h10H,1-8H2 |
| SMILES | OCCCCCCCCBr |
| IUPAC Name | 8-bromooctan-1-ol |
| Molecular Weight (g/mol) | 209.12 |
| Formula Weight | 208.0463 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction