accessibility menu, dialog, popup

UserName

CAS RN 505-66-8

NHHN

CAS RN 505-66-8

IUPAC Name: 1,4-diazepane
Synonyms: 1H-1,4-diazepine, hexahydro-
Molecular Weight (g/mol): 100.17
Molecular Formula: C5H12N2
InChi Key: FQUYSHZXSKYCSY-UHFFFAOYSA-N
SMILES: C1CNCCNC1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (5)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)