accessibility menu, dialog, popup

UserName

CH3SOOOONHOHOOH

CAS RN 501919-59-1

IUPAC Name: 2-hydroxy-4-{2-[(4-methylbenzenesulfonyl)oxy]acetamido}benzoic acid
Synonyms: 2-hydroxy-4-{2-[(4-methylbenzenesulfonyl)oxy]acetamido}benzoic acid
Molecular Weight (g/mol): 365.36
Molecular Formula: C16H15NO7S
InChi Key: HWNUSGNZBAISFM-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCC(=O)NC1=CC(O)=C(C=C1)C(O)=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (4)