accessibility menu, dialog, popup

UserName

CAS RN 4903-09-7

H3COOCl

CAS RN 4903-09-7

IUPAC Name: 3-chloro-4-methoxybenzaldehyde
Synonyms: 3-chloro-4-methoxybenzaldehyde
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: WYVGYYIZXPXHAZ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=O)C=C1Cl
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)