accessibility menu, dialog, popup

UserName

HOOOHHO

CAS RN 490-79-9

IUPAC Name: 2,5-dihydroxybenzoic acid
Synonyms: 2,5-dihydroxybenzoic acid gentisinic acid gentisinate gensigon gensigen kyselina gentisinova acide gentisique kyselina 2,5-dihydroxybenzoova dihydroxybenzoicacid acido 2,5-diidrossibenzoico
Molecular Weight (g/mol): 154.12
Molecular Formula: C7H6O4
InChi Key: WXTMDXOMEHJXQO-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(O)=CC=C1O
Molecular Weight (g/mol) 
  • (10)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (5)
  • (1)
Physical Form 
  • (3)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (10)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)