Learn More
2,5-Dihydroxybenzoic Acid, Sigma-Aldrich™
Specifications
Specifications
| CAS | 490-79-9 |
| Melting Point | 202°C to 206°C |
| MDL Number | MFCD00002460 |
| Quantity | 50 g |
| Synonym | 2,5-Dihydroxybenzoic acid, 2,5-Dihydroxybenzoic acid |
| Molecular Weight (g/mol) | 154.12 |
| Grade | Research |