accessibility menu, dialog, popup

UserName

CAS RN 4867-37-2

H3CSCl

CAS RN 4867-37-2

IUPAC Name: 1-chloro-3-(methylsulfanyl)benzene
Synonyms: 1-chloro-3-(methylsulfanyl)benzene
Molecular Weight (g/mol): 158.64
Molecular Formula: C7H7ClS
InChi Key: PTGSDZVASWKUHK-UHFFFAOYSA-N
SMILES: CSC1=CC=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results