missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorothioanisole, 99%, Thermo Scientific™

Product Code. 11367726
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367726 5 g 5g
11357726 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367726 Supplier Thermo Scientific Alfa Aesar Supplier No. L11346.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4867-37-2
Molecular Formula C7H7ClS
Molecular Weight (g/mol) 158.643
MDL Number MFCD00040127
InChI Key PTGSDZVASWKUHK-UHFFFAOYSA-N
Synonym 3-chlorothioanisole, 3-chlorophenyl methyl sulfide, 3-chloro thioanisole, 1-chloro-3-methylsulfanyl benzene, m-chlorothioanisole, 3-methylthio chlorobenzene, 3-cpms, 3-chlorothioanisol, pubchem10644, acmc-1bnc1
PubChem CID 97585
IUPAC Name 1-chloro-3-methylsulfanylbenzene
SMILES CSC1=CC(=CC=C1)Cl

Specifications

Density 1.206
Boiling Point 218°C
Flash Point 88°C (190°F)
Odor Unpleasant
Refractive Index 1.599
Quantity 5 g
Beilstein 1931485
Formula Weight 158.65
Percent Purity 99%
Chemical Name or Material 3-Chlorothioanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.