accessibility menu, dialog, popup

UserName

CAS RN 486-56-6

CH3NON

CAS RN 486-56-6

IUPAC Name: 1-methyl-5-(pyridin-3-yl)pyrrolidin-2-one
Synonyms: (-)-cotinine (S)-(-)-cotinine cotininum cotinine (substance) S-(-)-cotinine cotinina scotine (S)-cotinine
Molecular Weight (g/mol): 176.22
Molecular Formula: C10H12N2O
InChi Key: UIKROCXWUNQSPJ-UHFFFAOYNA-N
SMILES: CN1C(CCC1=O)C1=CC=CN=C1
Molecular Weight (g/mol) 
  • (2)
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)