accessibility menu, dialog, popup

UserName

H3CCH3CH3H3CH3C

CAS RN 481-21-0

IUPAC Name: 9a,11a-dimethyl-1-(6-methylheptan-2-yl)-hexadecahydro-1H-cyclopenta[a]phenanthrene
Synonyms: cholestane
Molecular Weight (g/mol): 372.68
Molecular Formula: C27H48
InChi Key: XIIAYQZJNBULGD-UHFFFAOYNA-N
SMILES: CC(C)CCCC(C)C1CCC2C3CCC4CCCCC4(C)C3CCC12C
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)