CAS RN 46022-05-3
CAS RN 46022-05-3
IUPAC Name:
(1R,2R)-cyclohexane-1,2-dicarboxylic acid
Synonyms:
(1R,2R)-cyclohexane-1,2-dicarboxylic acid
Molecular Weight (g/mol):
172.18
Molecular Formula:
C8H12O4
InChi Key:
QSAWQNUELGIYBC-PHDIDXHHSA-N
SMILES:
OC(=O)[C@@H]1CCCC[C@H]1C(O)=O
1
–
2
of
2
results
1
(1R,2R)-Cyclohexane-1,2-dicarboxylic acid, 98+%
CAS: 46022-05-3 Molecular Formula: C8H12O4 Molecular Weight (g/mol): 172.18 MDL Number: MFCD00062973 InChI Key: QSAWQNUELGIYBC-PHDIDXHHSA-N Synonym: 1r,2r-cyclohexane-1,2-dicarboxylic acid,trans-1,2-cyclohexanedicarboxylic acid,trans-cyclohexane-1,2-dicarboxylic acid,1r,2r---1,2-cyclohexanedicarboxylic acid,1r,2r-1,2-cyclohexanedicarboxylic acid,1,2-cyclohexanedicarboxylic acid, 1r,2r-rel,1,2-cyclohexanedicarboxylic acid, 1r,2r,1r.2r-cyclohexane-1,2-dicarboxylicacid,1,2-cyclohexanedicarboxylic acid, trans,pubchem19603 PubChem CID: 720895 IUPAC Name: (1R,2R)-cyclohexane-1,2-dicarboxylic acid SMILES: C1CCC(C(C1)C(=O)O)C(=O)O