Learn More
(R,R)-1,2-Cyclohexanedicarboxylic Acid, TRC
Specifications
Specifications
| CAS | 46022-05-3 |
| Molecular Formula | C8H12O4 |
| Quantity | 1 g |
| Synonym | (1R-trans)-1,2-Cyclohexanedicarboxylic Acid, (1R,2R)-Cyclohexane-1,2-dicarboxylic Acid; |
| InChI Formula | InChI=1S/C8H12O4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h5-6H,1-4H2,(H,9,10)(H,11,12)/t5-,6-/m1/s1 |
| SMILES | OC(=O)[C@@H]1CCCC[C@H]1C(=O)O |
| IUPAC Name | (1R,2R)-cyclohexane-1,2-dicarboxylic acid |
| Molecular Weight (g/mol) | 172.18 |
| Formula Weight | 172.0736 |
| Chemical Name or Material | (R,R)-1,2-Cyclohexanedicarboxylic Acid |