CAS RN 452-10-8
IUPAC Name:
2,4-difluoro-1-methoxybenzene
Synonyms:
2,4-difluoro-1-methoxybenzene
Molecular Weight (g/mol):
144.12
Molecular Formula:
C7H6F2O
InChi Key:
CRMJLJFDPNJIQA-UHFFFAOYSA-N
SMILES:
COC1=CC=C(F)C=C1F
1
–
1
of
1
results
1
2,4-Difluoroanisole, 98%, Thermo Scientific™
CAS: 452-10-8 Molecular Formula: C7H6F2O Molecular Weight (g/mol): 144.12 MDL Number: MFCD00010731 InChI Key: CRMJLJFDPNJIQA-UHFFFAOYSA-N Synonym: 2,4-difluoroanisole,2,4-difluoroanilsole,2,4-difluoromethoxy benzene,benzene, 2,4-difluoro-1-methoxy,pubchem3455,2,4-difluoro anisole,ksc493m8b,2,4-difluoro-1-methoxy-benzene,# PubChem CID: 136293 IUPAC Name: 2,4-difluoro-1-methoxybenzene SMILES: COC1=CC=C(F)C=C1F