accessibility menu, dialog, popup

UserName

H3COHNBrFNNONH3C

CAS RN 443913-73-3

IUPAC Name: N-(4-bromo-2-fluorophenyl)-6-methoxy-7-[(1-methylpiperidin-4-yl)methoxy]quinazolin-4-amine
Synonyms: vandetanib zactima C22H24BrFN4O2 vande-tanib
Molecular Weight (g/mol): 475.36
Molecular Formula: C22H24BrFN4O2
InChi Key: UHTHHESEBZOYNR-UHFFFAOYSA-N
SMILES: COC1=C(OCC2CCN(C)CC2)C=C2N=CN=C(NC3=CC=C(Br)C=C3F)C2=C1
Molecular Weight (g/mol) 
  • (7)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (5)
  • (2)
  • (7)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)