accessibility menu, dialog, popup

UserName

CAS RN 42835-25-6

CH3FONOHO

CAS RN 42835-25-6

IUPAC Name: 7-fluoro-12-methyl-4-oxo-1-azatricyclo[7.3.1.0⁵,¹³]trideca-2,5,7,9(13)-tetraene-3-carboxylic acid
Synonyms: flumequine substipharm brand of flumequine 氟甲喹 flumequinum FLM apurone flumigal flumisol flumiquil imequyl
Molecular Weight (g/mol): 261.25
Molecular Formula: C14H12FNO3
InChi Key: DPSPPJIUMHPXMA-UHFFFAOYNA-N
SMILES: CC1CCC2=C3N1C=C(C(O)=O)C(=O)C3=CC(F)=C2
Molecular Weight (g/mol) 
  • (7)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (2)
  • (1)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (7)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)