missing translation for 'onlineSavingsMsg'
Learn More

Flumequine, TRC

Product Code. 30004396
Change view
Click to view available options
Quantity:
10 mg
50 mg
1 g
Unit Size:
10mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004396 50 mg 50mg
30003646 1 g 1g
17962515 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004396 Supplier Toronto Research Chemicals Supplier No. TRCF44500050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42835-25-6
Molecular Formula C14 H12 F N O3
Molecular Weight (g/mol) 261.25
Synonym Flumequine, (RS)-9-Fluoro-5-methyl-1-oxo-6,7-dihydro-1H,5H-benzo[i,j]quinolizine-2-carboxylic acid, 1H,5H-Benzo[ij]quinolizine-2-carboxylic acid, 9-fluoro-6,7-dihydro-5-methyl-1-oxo-, 9-Fluoro-6,7-dihydro-5-methyl-1-oxo-1H,5H-benzo[ij]quinolizine-2-carboxylic acid (ACI), (±)-Flumequine, Apurone, Fantacin, Firestop, Flumequine, Flumigal, Flumiquil, Flumisol, Flumix, Imequyl, R 802, R 802 (bactericide)
IUPAC Name 7-fluoro-12-methyl-4-oxo-1-azatricyclo[7.3.1.05,13]trideca-2,5,7,9(13)-tetraene-3-carboxylic acid
SMILES CC1CCc2cc(F)cc3C(=O)C(=CN1c23)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H12FNO3/c1-7-2-3-8-4-9(15)5-10-12(8)16(7)6-11(13(10)17)14(18)19/h4-7H,2-3H2,1H3,(H,18,19)
Formula Weight 261.0801
Chemical Name or Material Flumequine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.