missing translation for 'onlineSavingsMsg'
Learn More

Flumequine 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30313860
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30313860 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313860 Supplier Dr. Ehrenstorfer Supplier No. DREA13718000AL1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Flumequine
CAS 42835-25-6
Analyte or Component Names Flumequine
Shipping Condition +20°C
Molecular Formula C14 H12 F N O3
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Flumequine, (RS)-9-Fluoro-5-methyl-1-oxo-6,7-dihydro-1H,5H-benzo[i,j]quinolizine-2-carboxylic acid
InChI Formula InChI=1S/C14H12FNO3/c1-7-2-3-8-4-9(15)5-10-12(8)16(7)6-11(13(10)17)14(18)19/h4-7H,2-3H2,1H3,(H,18,19)
SMILES CC1CCc2cc(F)cc3C(=O)C(=CN1c23)C(=O)O
IUPAC Name (RS)-9-Fluoro-5-methyl-1-oxo-6,7-dihydro-1H,5H-benzo[i,j]quinolizine-2-carboxylic acid
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 261.25
Quantity 1 mL
Formula Weight 261.0801 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.