CAS RN 4276-49-7
CAS RN 4276-49-7
IUPAC Name:
1-bromoicosane
Synonyms:
1-bromoeicosane
Molecular Weight (g/mol):
361.45
Molecular Formula:
C20H41Br
InChi Key:
CZASMUMJSKOHFJ-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCCCCBr
1
–
3
of
3
results
1
1-Bromoeicosane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-Bromoeicosane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1-Bromoeicosane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.