missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoeicosane, TRC

Product Code. 30345885
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30345885 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30345885 Supplier Toronto Research Chemicals Supplier No. TRCB678838250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Bromoeicosane
CAS 4276-49-7
Molecular Formula C20H41Br
Quantity 250 mg
Synonym 1-Bromoicosane, 1-Ecosyl Bromide, Eicosyl Bromide, NSC 87634, n-Eicosyl Bromide
InChI Formula InChI=1S/C20H41Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21/h2-20H2,1H3
SMILES CCCCCCCCCCCCCCCCCCCCBr
IUPAC Name 1-bromoicosane
Molecular Weight (g/mol) 361.44
Formula Weight 360.2392
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.