accessibility menu, dialog, popup

UserName

H3CNHOCl

CAS RN 42276-57-3

IUPAC Name: 4-chloro-N-(2-ethylphenyl)butanamide
Synonyms: 4-chloro-N-(2-ethylphenyl)butanamide
Molecular Weight (g/mol): 225.72
Molecular Formula: C12H16ClNO
InChi Key: VAWRRNXJFXHSSI-UHFFFAOYSA-N
SMILES: CCC1=CC=CC=C1NC(=O)CCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results