accessibility menu, dialog, popup

UserName

H3CNNH2N

CAS RN 41926-06-1

IUPAC Name: 1-methyl-1H-indazol-7-amine
Synonyms: 1-methylindazol-7-amine
Molecular Weight (g/mol): 147.18
Molecular Formula: C8H9N3
InChi Key: FKQDTYKVWIDGNT-UHFFFAOYSA-N
SMILES: CN1N=CC2=CC=CC(N)=C12
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results