accessibility menu, dialog, popup

UserName

CAS RN 41443-28-1

OONNN

CAS RN 41443-28-1

IUPAC Name: 1H-[1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one
Synonyms: [1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one
Molecular Weight (g/mol): 187.16
Molecular Formula: C9H5N3O2
InChi Key: LZMHWZHOZLVYDL-UHFFFAOYSA-N
SMILES: O=C1ON=C2C=NC3=CC=CC=C3N12
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)