CAS RN 40800-90-6
CAS RN 40800-90-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-(Methoxymethylene)-2(3H)-benzofuranone, TRC
CAS: 40800-90-6 Chemical Name or Material: 3-(Methoxymethylene)-2(3H)-benzofuranone Formula Weight: 176.0473 InChI Formula: InChI=1S/C10H8O3/c1-12-6-8-7-4-2-3-5-9(7)13-10(8)11/h2-6H,1H3/b8-6- IUPAC Name: (3Z)-3-(methoxymethylidene)-1-benzofuran-2-one Molecular Formula: C10H8O3 Molecular Weight (g/mol): 176.17 Recommended Storage: -20°C SMILES: CO\C=C\1/C(=O)Oc2ccccc12