accessibility menu, dialog, popup

UserName

OHNClClClHNSHNN(E/Z)

CAS RN 405060-95-9

IUPAC Name: 3-phenyl-N-(2,2,2-trichloro-1-{[(quinolin-8-yl)carbamothioyl]amino}ethyl)prop-2-enamide
Synonyms: salubrinal
Molecular Weight (g/mol): 479.8
Molecular Formula: C21H17Cl3N4OS
InChi Key: LCOIAYJMPKXARU-UHFFFAOYNA-N
SMILES: ClC(Cl)(Cl)C(NC(=S)NC1=C2N=CC=CC2=CC=C1)NC(=O)C=CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (8)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Color 
  • (5)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results