accessibility menu, dialog, popup

UserName

CAS RN 403842-84-2

H3CNNCH3FFFSOOO

CAS RN 403842-84-2

IUPAC Name: 3-methyl-1-octyl-1H-imidazol-3-ium trifluoromethanesulfonate
Synonyms: 1-octyl-3-methylimidazolium triflate
Molecular Weight (g/mol): 344.39
Molecular Formula: C13H23F3N2O3S
InChi Key: BCVMAYRUJBXMAP-UHFFFAOYSA-M
SMILES: [O-]S(=O)(=O)C(F)(F)F.CCCCCCCCN1C=C[N+](C)=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results