accessibility menu, dialog, popup

UserName

HOOFN

CAS RN 403-45-2

IUPAC Name: 6-fluoropyridine-3-carboxylic acid
Synonyms: 6-fluoropyridine-3-carboxylic acid
Molecular Weight (g/mol): 141.1
Molecular Formula: C6H4FNO2
InChi Key: UJDLCTNVHJEBDG-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(F)N=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results