CAS RN 40150-92-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1-(4-Isobutylphenyl)ethanol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Alpha-(4-Isobutylphenyl)ethanol, TRC
CAS: 40150-92-3 Molecular Formula: C12 H18 O Molecular Weight (g/mol): 178.27 Synonym: 1-(4-Isobutylphenyl)ethanol IUPAC Name: 1-[4-(2-methylpropyl)phenyl]ethanol SMILES: CC(C)Cc1ccc(cc1)C(C)O