Learn More
1-(4-Isobutylphenyl)ethanol, Mikromol™
Product Code. 17964286
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17964286 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17964286 Supplier Mikromol Supplier No. MM0002.50
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(4-Isobutylphenyl)ethanol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-(4-Isobutylphenyl)ethanol
CAS 40150-92-3
Analyte or Component Names 1-(4-Isobutylphenyl)ethanol
Shipping Condition Ice pack (-20°C)
Molecular Formula C12 H18 O
Certifications/Compliance ISO 17025
Synonym 1-(4-Isobutylphenyl)ethanol
InChI Formula InChI=1S/C12H18O/c1-9(2)8-11-4-6-12(7-5-11)10(3)13/h4-7,9-10,13H,8H2,1-3H3
SMILES CC(C)Cc1ccc(cc1)C(C)O
IUPAC Name 1-[4-(2-methylpropyl)phenyl]ethanol
Molecular Weight (g/mol) 178.27
Quantity 100 mg
Formula Weight 178.1358 g/mol