accessibility menu, dialog, popup

UserName

H3COFH3C

CAS RN 399-54-2

IUPAC Name: 4-fluoro-1-methoxy-2-methylbenzene
Synonyms: 4-fluoro-1-methoxy-2-methylbenzene
Molecular Weight (g/mol): 140.16
Molecular Formula: C8H9FO
InChi Key: QXOBYWRKNIDHJG-UHFFFAOYSA-N
SMILES: COC1=CC=C(F)C=C1C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results