accessibility menu, dialog, popup

UserName

CAS RN 399-31-5

CH3ONHF

CAS RN 399-31-5

IUPAC Name: N-(2-fluorophenyl)acetamide
Synonyms: 2-fluoroacetanilide
Molecular Weight (g/mol): 153.16
Molecular Formula: C8H8FNO
InChi Key: AUZPZBPZWHEIDY-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC=C1F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results