accessibility menu, dialog, popup

UserName

ONBr

CAS RN 39489-67-3

IUPAC Name: 2-(4-bromophenoxy)acetonitrile
Synonyms: 2-(4-bromophenoxy)acetonitrile
Molecular Weight (g/mol): 212.05
Molecular Formula: C8H6BrNO
InChi Key: PMDDXFGYBKIPKR-UHFFFAOYSA-N
SMILES: BrC1=CC=C(OCC#N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results