accessibility menu, dialog, popup

UserName

FFFClNH2

CAS RN 39226-96-5

IUPAC Name: 1-[2-chloro-3-(trifluoromethyl)phenyl]methanamine
Synonyms: 1-[2-chloro-3-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol): 209.6
Molecular Formula: C8H7ClF3N
InChi Key: GRFVQNWTQVWNBY-UHFFFAOYSA-N
SMILES: NCC1=C(Cl)C(=CC=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results