accessibility menu, dialog, popup

UserName

H3CSNNNHNOONH

CAS RN 39069-52-8

IUPAC Name: N4,N6-dicyclopentyl-2-(methylsulfanyl)-5-nitropyrimidine-4,6-diamine
Synonyms: N4,N6-dicyclopentyl-2-(methylsulfanyl)-5-nitropyrimidine-4,6-diamine
Molecular Weight (g/mol): 337.44
Molecular Formula: C15H23N5O2S
InChi Key: GSGVDKOCBKBMGG-UHFFFAOYSA-N
SMILES: CSC1=NC(NC2CCCC2)=C(C(NC2CCCC2)=N1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results