CAS RN 38520-57-9
CAS RN 38520-57-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Tocris Bioscience™ N-Acetylcysteine amide
Glutathione (GSH) precursor; neuroprotective
2
N-Acetylcysteine amide, MedChemExpress
MedChemExpress N-Acetylcysteine amide is a cell membranes and blood brain barrier permeant thiol antioxidant and neuroprotective agent, reduces ROS production.
3
4