missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetylcysteine amide, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
N-Acetylcysteine amide is a cell membranes and blood brain barrier permeant thiol antioxidant and neuroprotective agent, reduces ROS production.
Specifications
Specifications
| Chemical Name or Material | N-Acetylcysteine amide |
| Physical Form | Solid |
| CAS | 38520-57-9 |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C5H10N2O2S |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | H2O : 200 mg/mL (1232.97 mM; Need ultrasonic) ∣DMSO : ≥ 100 mg/mL (616.48 mM) |
| SMILES | CC(N[C@@H](CS)C(N)=O)=O |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction