accessibility menu, dialog, popup

UserName

CH3H3COONOOCH3H3CNHO

CAS RN 37558-29-5

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 1-hydroxy-2,2,5,5-tetramethyl-2,5-dihydro-1H-pyrrole-3-carboxylate
Synonyms: 2,5-dioxopyrrolidin-1-yl 1-hydroxy-2,2,5,5-tetramethylpyrrole-3-carboxylate
Molecular Weight (g/mol): 282.3
Molecular Formula: C13H18N2O5
InChi Key: VDSVTMQVXNIMMJ-UHFFFAOYSA-N
SMILES: CC1(C)C=C(C(=O)ON2C(=O)CCC2=O)C(C)(C)N1O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)