CAS RN 37158-47-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Clenpenterol Hydrochloride, TRC
CAS: 37158-47-7 Molecular Formula: C13 H20 Cl2 N2 O . Cl H Molecular Weight (g/mol): 327.68 Synonym: Benzenemethanol, 4-amino-3,5-dichloro-α-[[(1,1-dimethylpropyl)amino]methyl]-, hydrochloride (1:1),Benzenemethanol, 4-amino-3,5-dichloro-α-[[(1,1-dimethylpropyl)amino]methyl]-, monohydrochloride (9CI),4-Amino-3,5-dichloro-α-[[(1,1-dimethylpropyl)amino]methyl]benzenemethanol hydrochloride (1:1),Clenpenterol hydrochloride IUPAC Name: 1-(4-amino-3,5-dichlorophenyl)-2-(2-methylbutan-2-ylamino)ethanol;hydrochloride SMILES: Cl.CCC(C)(C)NCC(O)c1cc(Cl)c(N)c(Cl)c1