accessibility menu, dialog, popup

UserName

NNNNNN

CAS RN 3682-35-7

IUPAC Name: tris(pyridin-2-yl)-1,3,5-triazine
Synonyms: tris(pyridin-2-yl)-1,3,5-triazine
Molecular Weight (g/mol): 312.34
Molecular Formula: C18H12N6
InChi Key: KMVWNDHKTPHDMT-UHFFFAOYSA-N
SMILES: C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1
Quantity 
  • (1)
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (1)
  • (4)