missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tripyridyl-S-triazine, 101.0%, MP Biomedicals™

Product Code. 11413531 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413531 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11413531 Supplier MP Biomedicals Supplier No. 0210312905
  • ONLINE EXCLUSIVE PRICE
    161.00 EUR
    153.30 EUR / 5g
    Save 7.70 EUR
    4% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 3682-35-7
Color Brown
Assay Percent Range 1.01
Molecular Formula C18H12N6
MDL Number MFCD00006045
Quantity 5 g
Synonym 2,4,6-tripyridyl-s-triazine, 2,4,6-tris 2-pyridyl-s-triazine, 2,4,6-tri 2-pyridyl-1,3,5-triazine, terpyridyl-s-triazine, 2,4,6-tri 2-pyridyl-s-triazine, tptz iron reagent, 1,3,5-triazine, 2,4,6-tri-2-pyridinyl, tri-2-pyridyl-s-triazine, s-triazine, tri-2-pyridyl, 2,4,6-tris 2-pyridyl-1,3,5-triazine
InChI Key KMVWNDHKTPHDMT-UHFFFAOYSA-N
SMILES C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1
IUPAC Name tris(pyridin-2-yl)-1,3,5-triazine
Molecular Weight (g/mol) 312.34
PubChem CID 77258
Formula Weight 312.3
Physical Form Fine Powder
Chemical Name or Material 2, 4, 6-Tripyridyl-S-triazine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.