accessibility menu, dialog, popup

UserName

CH3CH3O(R)(R)HOO(R)(R)ONNONNH2NH

CAS RN 362-76-5

IUPAC Name: 9-[(3aR,4R,6R,6aR)-6-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-yl]-2-amino-6,9-dihydro-3H-purin-6-one
Synonyms: 9-[(3aR,4R,6R,6aR)-6-(hydroxymethyl)-2,2-dimethyl-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]-2-amino-3H-purin-6-one
Molecular Weight (g/mol): 323.31
Molecular Formula: C13H17N5O5
InChi Key: XKPDAYWPKILAMO-IOSLPCCCSA-N
SMILES: CC1(C)O[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O1)N1C=NC2=C1NC(N)=NC2=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)