missing translation for 'onlineSavingsMsg'
Learn More

2',3'-O-Isopropylidene-Guanosine, 98%, MP Biomedicals™

Product Code. 11338959 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 11338959

Brand: MP Biomedicals 0210209901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 362-76-5
Color White
Quantity 1 g
Physical Form Powder
Assay Percent Range 98%
Molecular Formula C13H17N5O5
MDL Number MFCD00057081
Synonym 2',3'-o-isopropylideneguanosine, 2-amino-9-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl-1h-purin-6 9h-one, 2',3'-o-1-methylethylidene guanosine, 2',3'-o-1-methylethylidene-guanosine, 9-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl-2-amino-3h-purin-6-one, 9-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl-2-amino-1h-purin-6-one, 2', 3'-isopropylideneguanosine, 2'-o,3'-o-isopropylideneguanosine, 2',3'-o-1-methylethyliden-guanosin, 2/',3/'-o-1-methylethylidene guanosine
InChI Key XKPDAYWPKILAMO-IOSLPCCCSA-N
SMILES CC1(C)O[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O1)N1C=NC2=C1NC(N)=NC2=O
IUPAC Name 9-[(3aR,4R,6R,6aR)-6-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-yl]-2-amino-6,9-dihydro-3H-purin-6-one
Molecular Weight (g/mol) 323.31
PubChem CID 6541409
Formula Weight 323.3
Chemical Name or Material 2', 3'-O-Isopropylidene-Guanosine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.