accessibility menu, dialog, popup

UserName

H3COOCH3H3CCH3

CAS RN 3549-23-3

IUPAC Name: methyl 2-(4-tert-butylphenyl)acetate
Synonyms: methyl 2-(4-tert-butylphenyl)acetate
Molecular Weight (g/mol): 206.29
Molecular Formula: C13H18O2
InChi Key: HXVTYMWVMVKVTF-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC=C(C=C1)C(C)(C)C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results