CAS RN 352431-15-3
IUPAC Name:
2-(4-chloro-2-methylphenoxy)propanoic acid
Synonyms:
mecoprop MCPP mecoprop-P
Molecular Weight (g/mol):
214.65
Molecular Formula:
C10H11ClO3
InChi Key:
WNTGYJSOUMFZEP-UHFFFAOYNA-N
SMILES:
CC(OC1=C(C)C=C(Cl)C=C1)C(O)=O
1
–
5
of
5
results
1
(±)-2-(4-Chloro-2-methylphenoxy-d3)propionic Acid, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
Mecoprop-d3, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Mecoprop-d3, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Mecoprop-d3, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.