missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mecoprop-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Mecoprop D3 (phenyl D3) |
| CAS | 352431-15-3 |
| Molecular Formula | C10 2H3 H8 Cl O3 |
| Quantity | 5 mg |
| Synonym | Propanoic acid, 2-(4-chloro-2-methylphenoxy-2,3,5-d3)- (9CI), Mecoprop D3 (phenyl D3), Mecoprop-d3, (±)-2-(4-Chloro-2-methylphenoxy-d3)propionic Acid |
| InChI Formula | InChI=1S/C10H11ClO3/c1-6-5-8(11)3-4-9(6)14-7(2)10(12)13/h3-5,7H,1-2H3,(H,12,13)/i3D,4D,5D |
| SMILES | [2H]c1c([2H])c(OC(C)C(=O)O)c(C)c([2H])c1Cl |
| IUPAC Name | 2-(4-chloro-2,3,5-trideuterio-6-methylphenoxy)propanoic acid |
| Molecular Weight (g/mol) | 217.66 |
| Formula Weight | 217.0585 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?