accessibility menu, dialog, popup

UserName

NH2SOOBrCl

CAS RN 351003-59-3

IUPAC Name: 4-bromo-2-chlorobenzene-1-sulfonamide
Synonyms: 4-bromo-2-chlorobenzenesulfonamide
Molecular Weight (g/mol): 270.53
Molecular Formula: C6H5BrClNO2S
InChi Key: BFZLQNCZBMUSRS-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=C(Br)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results